Learn More
Thermo Scientific™ Remel™ CCFA Modified Agar (with cefoxitin, cycloserine, fructose)
Catalog No. r01268
This item is not returnable. View return policy
This item is not returnable. View return policy
Remel CCFA Modified Agar (with cefoxitin, cycloserine, fructose) is used for selective isolation and differentiation of Clostridium difficile.
Thermo Scientific™ Remel CCFA Modified Agar (with cefoxitin, cycloserine, fructose) is used for selective isolation and differentiation of Clostridium difficile.
TRUSTED_SUSTAINABILITY
Specifications
Description CCFA Modified Agar w/cefoxitin, cycloserine, fructose
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.