Learn More
Thermo Scientific™ Chocolate Agar w/Bacitracin
Catalog No. r01304
This item is not returnable. View return policy
This item is not returnable. View return policy
Isolate Haemophilus species from respiratory specimens containing mixed flora with this medium.
Isolate Haemophilus species from respiratory specimens containing mixed flora with Thermo Scientific™ Remel™ Chocolate Agar w/Bacitracin. Isolation and identification of Haemophilus spp is often hindered by overgrowth with commensal microbial flora. Hoving and Aandahl reported improved recovery of Haemophilus on Chocolate Agar with Bacitracin as compared to isolation on Sheep Blood Agar with a streak of Staphylococcus aureus.
Hemoglobin in the Chocolate Agar formulation provides the X factor, and GCHI Enrichment provides the V factor required for growth of fastidious organisms. The presence of corn-starch ensures that toxic metabolites produced Haemophilus by are absorbed. Bacitracin is a selective agent which is inhibitory to most strains of streptococci, staphylococci, Neiseeria spp., and Micococcus spp.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Chocolate Agar w/Bacitracin
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.