Learn More
Thermo Scientific™ Remel™ Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80
                    Click to view available options
                                Quantity:
                                100/Pk.
                                20/Pk.
                    Description
Thermo Scientific™ Remel Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80 is used for the cultivation of bacteria, yeast and fungi.
- Manufactured in a clean room environment to provide the highest level of sterility assurance with no compromise of growth characteristics
 - Each lot is tested to ensure consistent, reproducible results
 - Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
            Specifications
Specifications
| Description | Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80 | 
| Format | Contact - Single Bag | 
| Product Type | Agar | 
| Quantity | 20/Pk. | 
Not intended for IVD use.