Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dermatophyte Test Medium (DTM)

Catalog No. p-4522792
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
F - slant
Monoplate Deep Pour
This item is not returnable. View return policy
This item is not returnable. View return policy

Facilitate selective and differential isolation of pathogenic fungi (dermatophytes) from cutaneous sources using Dermatophyte Test Medium (DTM).

Facilitate selective and differential isolation of pathogenic fungi (dermatophytes) from cutaneous sources using Thermo Scientific™ Remel™ Dermatophyte Test Medium (DTM). This medium was formulated by Taplin et al. for use in locations where specialized training and microscopic examination is not available1. A pH indicator and three antimicrobial agents are added to the agar to provide a differential and selective medium for isolation of dermatophytes belonging to the genera Microsporum, Trichophyton, or Epidermophyton.

Contains a pH indicator.
TRUSTED_SUSTAINABILITY

Specifications

Description Dermatophyte Test Medium
Format F - slant
Product Type Microbiology Media
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.