Learn More
Thermo Scientific™ DNase Test Agar
Catalog No. r01375
This item is not returnable. View return policy
This item is not returnable. View return policy
Detect DNase activity in Staphylococci spp., enteric gram-negative bacilli and pseudomonads with DNase Test Agar.
Detect the deoxyribonuclease (DNase) activity in microorganisms with Thermo Scientific™ Remel™ DNase Test Agar.
In 1957, Jeffries et al. developed a medium for demonstrating deoxyribonuclease (DNase) activity of microorganisms possessing this extracellular enzyme.1 Such microorganisms depolymerize DNA in the medium resulting in a clear zone around the colonies when the plate is flooded with 1N hydrochloric acid (HCl). DNase Test Agar has been recommended for identification of a variety of bacteria including staphylococci, enteric gram-negative bacilli, and pseudomonads.2-5
This is a solid medium used to detect the deoxyribonuclease (DNase) activity in microorganisms.
  • Easy-to-interpret:
    • Positive test – after 1N HCl is added, a clear zone is formed around the growth line and the rest of the plate remain opaque.
    • Negative test – after 1N HCl is added, no clear zone is formed around the growth line and the entire plate is opaque.
  • Ready-to-use: convenience of prepared media.
DNase Test Agar contain casein and soy peptone that supply nitrogen, amino acids and peptides necessary for bacterial growth. Sodium chloride provide essential electrolytes which maintain osmotic equilibrium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description DNase Test Agar
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.