Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Eugonic Broth (Modified)

Catalog No. r07104
Click to view available options
Quantity:
100 x 10 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Cultivate a variety of microorganisms, such as Neisseria, Haemophilus and anaerobic bacteria with Eugonic Broth (Modified).

Cultivate a variety of microorganism, such as Neisseria, Haemophilus and anaerobic bacteria with Thermo Scientific™ Remel™ Eugonic Broth (Modified).

Eugonic Broth (Modified) was developed by Vera to obtain eugonic growth of fastidious organism, such as Neisseria, Haemophilus and Brucella1,2. Finegold recommended the addition of maltose, yeast extract, vitamin K and hemin to help in the recovery of anaerobe and fastidious organism from clinical specimen3. Campos and Spainhour recommended this broth for detecting bacteremia in pediatric patients4.

Eugonic Broth (Modified), is a liquid medium recommended for cultivation of microorganism such as Neisseria, Haemophilus and anaerobic bacteria. This broth is used to detect bacteremia in patients from whom only a small amount of blood can be collected.

  • Ready-to-use: Convenience of prepared media.
  • Easy-to-use: Inoculate the broth directly with clinical specimen immediately once it is received in the laboratory and incubate at 33-37º C for 72 hours.

Eugonic Broth (Modified) contains casein, meat and soy peptones that supply essential nutrients which support rapid growth of microorganisms. Dextrose provide ready source of energy. Maltose and yeast extract stimulate growth of anaerobic bacteria. GCHI Enrichment is a supplement which provide NAD, vitamins, amino acids, coenzymes, dextrose and ferric ions to promote growth of fastidious microorganisms that include Neisseria and Haemophilus. Hemin and vitamin K supply growth factors required by anaerobes. A small quantity of agar is added to enhance anaerobic conditions.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Eugonic Broth Modified
Format 16mm x 125mm Tube
Product Type Microbiology Media
Quantity 100 x 10 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.