Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial

Catalog No. p-4525726
Click to view available options
Volume (Metric):
15 mL
30 mL
This item is not returnable. View return policy

Catalog No. R21616

Supplier: Thermo Scientific™ R21616

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.

Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.

Caps are color coded to match industry standard colors and have an attached spork.
  • Broad range with long shelf life
  • Industry standard color-coded caps and labels
  • Easy to use spork cap for safe collection
  • Convenient packaging, including bulk kits
  • Multilingual patient collection instructions
  • Empty Vial is used for test procedures not compatible with fecal preservatives
  • Cary-Blair with Indicator contains 0.16% agar and is for preservation and recovery of enteric bacterial pathogens
TRUSTED_SUSTAINABILITY

Specifications

Product Type Fecal Transport Set
Description Cary-Blair/Empty Vial; Two-vial Sets
Packaging 2-Vial Set
Volume (Metric) 15 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.