Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Fluid D w/0.1% peptone, 0.1% Polysorbate 80

Catalog No. p-4523214
Click to view available options
Quantity:
10 x 100 mL
10 x 300 mL
10/Pk.
Format:
Borosilicate Bottle - 100 mL
Borosilicate Bottle - Septum
Borosilicate Bottle, Septum
SB - 300mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo Scientific™ Remel Fluid D with 0.1% peptone, 0.1% Polysorbate 80 is used as diluting or rinsing fluid for sterility testing.

Thermo Scientific™ Remel Fluid D with 0.1% peptone, 0.1% Polysorbate 80 is used as diluting or rinsing fluid for sterility testing.

TRUSTED_SUSTAINABILITY

Specifications

Description Fluid D
Format SB - 300mL
Product Type Buffer Solution
Quantity 10 x 300 mL

Not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.