Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GC Agar Base w/1% GCHI

Catalog No. r04030
Click to view available options
Format:
15mm x 100mm Plate
15mm x 150mm Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Recommended for antimicrobial susceptibility testing of Neisseria gonorrhoeae by the disc diffusion technique.

Perform Antimicrobial disc diffusion susceptibility testing of Neisseria gonorrhoea with Thermo Scientific™ Remel™ GC Agar Base with 1% GCHI.

In 1945, Johnston developed modified chocolate agar that allowed the growth of N. gonorrhoea in 24 hours rather than 48 hours1. Fast growth rate was traced to differences in gel strength of the media tested. Therefore, GC Agar Base w1% GCHI was developed with reduced agar content. In 1977, Thornsberry et al. used GC Agar Base to determine β-lactamase production by N. gonorrhoeae2. Jones et al. recommended adding a modified GCHI Enrichment into GC Agar Base3.

GC Agar Base w1% GCHI is a solid medium used for antimicrobial susceptibility testing of Neisseria gonorrhoea using the disc diffusion technique. The effect of penicillin is inhibited when pathogenic Neisseria produce β-lactamase. Gonococci with 10U penicillin disc zone diameters ≤ 19 mm are likely to be β-lactamase producing strains4.

  • Ready-to-use: Convenience of prepared media.
  • Easy-to-interpret:
    • Positive test- Penicillin-resistant organism with 10U penicillin disc zone diameter ≤ 19 mm.
    • Negative test- tiny or individual colonies showing no growth.
  • Meets CLSI standards.4

GC Agar Base w1% GCHI contains casein and meat peptones that supply corn starch to neutralize toxic fatty acids. The phosphate buffer controls pH changes that can affect organism survival. The GCHI Enrichment provides NAD, vitamins, amino acids, coenzymes, dextrose, ferric ions and other growth factors needed to cultivate Neisseria species.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description GC Agar Base
Format 15mm x 150mm Plate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.