Learn More
Thermo Scientific™ Remel™ GC Agar Base
Description
Remel GC Agar Base is used with hemoglobin and GCHI enrichment for the isolation and cultivation of Neisseria/gonorrhoeae and other fastidious microorganisms.
Specifications
Specifications
| Description | GC Agar Base |
| Form | Powder |
| Media Type | GC Agar Base |
| Product Type | Nutrient Agar |
| Quantity | 2.5 kg |
For Laboratory Use Only