Learn More
Thermo Scientific™ Remel™ GenBio ImmunoDOT™ Thyroid Autoimmunity Test Kit
Catalog No. r243912
Click to view available options
Quantity:
1 Vial
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Complete with assay strips, diluent, enhancer, conjugate, and developer.
Intended for use in physician offices and clinical laboratories
  • CLIA Complexity: Non-waived
  • For screening and detection of autoantibodies against human thyroglobulin and human thyroid peroxidase (microsome) in serum and heparinized whole blood
  • Qualitative results for 10μL serum or 20μL heparinized whole blood
  • Enzyme immunoassay (EIA) test
  • Reagents must be at room temperature (15° to 30°C, 59° to 86°F) before use; use within one hour of placement in the heated workstation
  • For in vitro diagnostic use
TRUSTED_SUSTAINABILITY
Specifications
Clia Complexity Non-waived
Type Thyroid Positive Control
For Use With (Application) Intended for use in physician offices and clinical laboratories
Includes Assay strips, diluent, enhancer, conjugate and developer
Quantity 1 Vial
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.