Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Haemophilus Test Medium Broth (HTM Broth)

Catalog No. p-4523863
Click to view available options
Quantity:
500 mL
20 x 10 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Use for antimicrobial minimal inhibitory concentration (MIC) determinations of Haemophilus spp.

Thermo Scientific™ Remel Haemophilus Test Medium Broth (HTM Broth) is for the antimicrobial minimal inhibitory concentration (MIC) determinations of Haemophilus spp.

TRUSTED_SUSTAINABILITY

Specifications

Description Haemophilus Test Medium Broth
Format 16mm x 125mm Tube
Product Type Microbiology Media
Quantity 20 x 10 mL

REF R112380 not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.