Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HI Broth (5mL)

Catalog No. p-4523867
Click to view available options
Quantity:
10 x 5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Cultivate and isolate fastidious and nonfastidious microorganisms using Heart Infusion Broth.

Cultivate and isolate fastidious and nonfastidious microorganisms using Thermo Scientific™ Remel™ Heart Infusion Broth. In the first media meat infusions were included for the cultivation of bacteria1. Huntoon prepared a medium using fresh beef heart and peptone which was known as Heart Infusion Broth. He demonstrated that without the addition of enrichment such as animal blood, it could be used to support the growth of nutritionally fastidious microorganisms.

  • Used for mass cultivation of bacteria required in vaccine preparation and animal blood can also be added to enable determination of hemolytic reactions by organisms2,3.
  • Ready to use convenience of prepared media.

This medium contains beef heart infusion and casein peptone that supply nitrogenous compounds and amino acids necessary for the growth of nutritionally fastidious bacteria. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium. Yeast extract is a source of energy and to enhance the bacterial growth it supplies B vitamins.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF.
Description Heart Infusion Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 10 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.