Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Inhibitory Mold Agar

Catalog No. p-4523438
Click to view available options
Quantity:
10 x 26 mL
10 x 35 mL
100/Pk.
Format:
20mm x 113mm Tube
Monoplate - 26mL
Monoplate - 35mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Primarily isolate and cultivate Histoplasma capsulatum, dermatophytes, and other pathogenic fungi using Inhibitory Mold Agar (w/chloramphenicol).

Thermo Scientific™ Remel™ Inhibitory Mold Agar (w/chloramphenicol) is used to primarily isolate and cultivate Histoplasma capsulatum, dermatophytes, and other pathogenic fungi. Ulrich formulated Inhibitory Mold Agar as a general-purpose medium for cultivation of fungi1. It is also recommended for selective isolation of pathogenic yeasts and molds such as Cryptococcus neoformans, H. capsulatum, and Trichophyton spp. from clinical specimens contaminated with commensal microbial flora2,3.

Inhibitory Mold Agar primarily supports isolation and cultivation of dermatophytes and other pathogenic fungi. Inhibitory Mold Agar contains casein and meat peptones that supply nitrogenous compounds, amino acids, and peptides necessary for the growth of fungi. Dextrose, starch, and dextrin are ready sources of energy. Sodium chloride and metallic salts are essential electrolytes that help to maintains osmotic equilibrium. Yeast extract is a source of vitamins.

  • Ready to use - Solid media
  • Recommended for selective isolation of pathogenic yeasts and molds such as Cryptococcus neoformans, capsulatum, and Trichophyton spp. from clinical specimens contaminated with commensal microbial flora2,3
  • Easy to read - Examine plates for fungal colonies that exhibit typical color and morphology
  • Selective media - Chloramphenicol inhibits many gram-positive and gram-negative bacteria while gentamicin, ciprofloxacin, and vancomycin inhibit commensal microbial flora
TRUSTED_SUSTAINABILITY

Specifications

Description Inhibitory Mold Agar
Format Monoplate - 26mL
Product Type Agar
Quantity 10 x 26 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.