Learn More
Thermo Scientific™ Inhibitory Mold Agar with Gentamicin and Chloramphenicol
Catalog No. r01506
This item is not returnable. View return policy
This item is not returnable. View return policy
Isolate and cultivate Histoplasma capsulatum, dermatophytes, and other pathogenic fungi with this selective, ready-to-use medium.
Isolate and cultivate Histoplasma capsulatum, dermatophytes, and other pathogenic fungi with Thermo Scientific™ Remel™ Inhibitory Mold Agar with Gentamicin and Chloramphenicol. Inhibitory Mold Agar was formulated by Ulrich as a general purpose medium for cultivation of fungi1. It is also recommended for selective isolation of pathogenic yeasts and molds (e.g. Cryptococcus neoformans, Histoplasma capulatum, and Trichophyton spp) from clinical specimens contaminated with commensal microbial flora2, 3.
Use Inhibitory Mold Agar with Gentamicin and Chloramphenicol for isolation and cultivation of Histoplasma capsulatum, dermatophytes, and other pathogenic fungi.
  • Selective: gentamicin and chloramphenicol act as selective agents. Chloramphenicol is a broad spectrum antibiotic which inhibits many Gram-positive and Gram-negative bacteria while gentamicin inhibits commensal microbial flora.
  • Ready-to-use: convenience of prepared media.
Casein and meat peptones supply nitrogenous compounds, amino acids, and peptides necessary for the growth of fungi. Dextrose, starch, and dextrin are ready sources of energy. Sodium chloride and metallic salts are essential electrolytes and help to maintain osmotic equilibrium. Yeast extract is a source of vitamins.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Inhibitory Mold Agar w/Gentamicin
Format Monoplate Deep Pour
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.