Learn More
Thermo Scientific™ JEMBEC System w/Thayer Martin Agar Modified (w/nystatin, colistin, vancomycin, trimethoprim)
Description
This medium is used to isolate Neisseria gonorrhoeae and Neisseria meningitidis.
Isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel JEMBEC System w/Thayer Martin Agar Modified (w/nystatin, colistin, vancomycin, trimethoprim). Johnston introduced a medium to grow N. gonorrhoeae in 24 hours1 and this was the advent of GC Agar Base. Thayer and Martin formulated a selective medium containing antibiotics to isolate N. gonorrhoeae and N. meningitidis 2. Various studies have demonstrated improved recovery for pathogenic Neisseria spp. using selective media3-6.
Initially the antibiotics ristocetin and polymyxin B were used and replaced with vancomycin, colistimethate, and nystatin7. The medium contains dextrose that enhances growth of gonococci.
- Ready-to-use – Convenience of prepared medium
- Selective media –
- Nystatin suppresses Candida albicans
- Colistin inhibits most gram-negative bacilli
- Vancomycin inhibits gram-positive cocci
- Trimethoprim lactate suppresses swarming of Proteus spp.
Thayer and Martin Agar (Improved) contains hemoglobin, which provides the X factor (hemin), and GCHI Enrichment that provides the V factor, vitamins, amino acids, coenzymes, and dextrose. The tablet provided in the JEMBEC™ and Pill Pocket systems comes in contact with the water vapor present in the medium and liberates carbon dioxide into the enclosed pack.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
Description | Modified; W/nystatin, colistin, vancomycin, trimethoprim |
Format | JEMBEC Plate |
Product Type | Agar |
Quantity | 10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.