Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Jordans Tartrate Agar (5mL Butt)

Catalog No. R061202 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061202 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R061202 Supplier Thermo Scientific™ Supplier No. R061202
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate Enterobacteriaceae based on tartrate utilization using Jordans Tartrate Agar.

Differentiate Enterobacteriaceae based on tartrate utilization using Thermo Scientific™ Remel™ Jordans Tartrate Agar. In 1928, Jordan and Harmon developed a medium to detect sodium tartrate fermentation by certain strains of Salmonella1. In 1957, Ellis et al. demonstrated that to differentiate Salmonella and Arizona, organic acids such as D-tartrate were useful2.

This agar medium differentiates Enterobacteriaceae based on tartrate utilization. This medium contains peptones that supply nitrogenous compounds and amino acids necessary for the growth of enteric gram-negative bacilli.

  • Ready to use – Convenience of prepared media.
  • Differential media - Sodium tartrate is a differential agent which is fermented by some enteric gram-negative bacilli
  • Easy to interpret -
    • Positive test - Yellow color development in the lower portion of the tube; the surface zone remains red
    • Negative test - No color change; agar remains alkaline with a red color throughout the tube

Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium. The acid produced results in a pH change causing the phenol indicator to change from red to yellow in the lower portion of the tube.

TRUSTED_SUSTAINABILITY

Specifications

Description Jordans Tartrate Agar
Format 15mm x 103mm Tube - butt
Product Type Agar
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.