Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Lactose Broth

Catalog No. r112401
Click to view available options
Format:
PB - 990mL
This item is not returnable. View return policy

Catalog No. R112401

Supplier: Thermo Scientific™ R112401

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel Lactose Broth is used for detection of coliform bacteria in water, foods, and dairy products according to standard methods.

Thermo Scientific™ Remel Lactose Broth is used for detection of coliform bacteria in water, foods, and dairy products according to standard methods.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, EPA, USDA, USP.
Description Lactose Broth
Format PB - 990mL
Product Type Microbiology Media
Quantity 10 x 90 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.