Learn More
Thermo Scientific™ MacConkey Sorbitol Agar with Cefixime and Tellurite
GSA_VA
Catalog No. R110241 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R110241 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R110241 Supplier Thermo Scientific™ Supplier No. R110241
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selectively detect sorbitol-negative Escherichia coli O157:H7 (VTEC) with MacConkey Sorbitol Agar with Cefixime and Tellurite.
Selectively detect sorbitol-negative Escherichia coli O157:H7 (VTEC) with Thermo Scientific™ Remel™ MacConkey Sorbitol Agar with Cefixime and Tellurite. The medium is recommended by Food and Drug Administration Center for Food Safety and Applied Nutrition (FDA/CFSAN) and the American Public Health Association (APHA) for selective isolation of VTEC from food samples6,7.
Verotoxin-producing Escherichia coli (VTEC), which includes serotype O157, is the fourth leading cause of diarrheal diseases in the United States4. This serotype of E. coli produces a verotoxin and is indistinguishable on the routine isolation media from those of non-VTEC E.coli. E. coli O157 is recognized as a cause of haemorrhagic colitis, an illness characterized by bloody diarrhoea and severe abdominal pain, and haemolytic uraemic syndrome (HUS)5 and as such, is a significant human pathogen.
Selectively detect sorbitol-negative Escherichia coli O157:H7 (VTEC) with MacConkey Sorbitol Agar with Cefixime and Tellurite.
  • Ease of differentiation: Due to the presence of D-sorbitol; sorbitol negative strains form colorless colonies while other E. coli strains that ferment sorbitol and form pink colonies.
  • Selective: Bile salts and crystal violet are selective agents which inhibit gram-positive organisms and enterococci, respectively. Cefixime and tellurite inhibit the growth of non-verotoxin producing E.coli and other non-sorbitol fermenting bacteria.
  • Recommended as a simple, inexpensive, rapid and reliable means of screening for E. coli O157
  • Recommended by FDA, Center for Food Safety and Applied Nutrition (FDA/CFSAN) and theAPHA for selective isolation of VTEC from food samples6,7.
In 1986, March and Ratnam introduced MacConkey Agar with sorbitol for the isolation of this serotype as unlike other serotypes VTEC does not ferment sorbitol1. In 1991, Chapman modified this medium by adding cefixime to specifically inhibit Proteus spp2. Zadik further modified it by adding potassium tellurite and found that it more effectively inhibited the growth of non-VTEC strains and increased isolation of E. coli O1573.
This medium contains peptones that provides nitrogenous compounds and amino acids essential for bacterial growth. Bile salts and crystal violet are selective agents which inhibit gram-positive organisms. Cefixime and tellurite inhibit the growth of non-verotoxin producing E.coli and other non-sorbitol fermenting bacteria. Lactose is substituted with D-sorbitol. Verotoxin- producing strains of E.coli ferment lactose but not sorbitol and form colorless colonies.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description MacConkey Agar w/Sorbitol, Cefixime, Tellurite
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.