Learn More
Thermo Scientific™ Remel™ MacConkey Agar
Catalog No. R453806
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453806 10 kg
R453802 500 g
R453804 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R453806 Supplier Thermo Scientific™ Supplier No. R453806
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
TRUSTED_SUSTAINABILITY
Specifications
Description MacConkey Agar
Form Powder
Media Type MacConkey Agar
Product Type Nutrient Agar
Quantity 10 kg