Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MacConkey Broth

Catalog No. r112454
Click to view available options
Format:
D - 5mL
PB - 100mL
WMB - 100mL

Selectively and differentially isolate enteric gram-negative bacilli using MacConkey Broth.

Selectively and differentially isolate enteric gram-negative bacilli using Thermo Scientific™ Remel™ MacConkey Broth. MacConkey Broth is a modification of the original bile salt broth first described by MacConkey in 1900, which contained litmus as an indicator and sodium taurocholate to inhibit gram-positive organisms1. In further testing, MacConkey recommended replacing litmus with neutral red2. In 1953, Childs and Allen replaced neutral red with bromcresol purple, a less inhibitory indicator3.

Also used in broth method for ProSpecT Microplate Assay, Part No. R2474048 and Part No. R2474096.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP
Description MacConkey Broth
Format WMB - 100mL
Product Type Microbiology Media
Quantity 10 x 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.