Learn More
Thermo Scientific™ McClung Toabe Agar
GSA_VA
Catalog No. R01056 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01056 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01056 Supplier Thermo Scientific™ Supplier No. R01056
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selectively isolate and differentiate Clostridium spp. from clinical specimens using McClung-Toabe Agar.
Selectively isolate and differentiate Clostridium spp. using Thermo Scientific™ Remel™ McClung-Toabe Agar. In 1947, McClung and Toabe developed this differential plating medium to detect lecithinase and lipase production by Clostridium spp1. This agar was further modified by Dowell and Hawkins by adding neomycin to selectively isolate and differentiate certain obligately anaerobic bacteria such as Clostridium perfringens2.
Use McClung-Toabe Agar for selectively isolate and differentiate Clostridium spp.
  • Selective – Neomycin inhibits many gram-positive and gram-negative organisms
  • Easy-to-interpret – Based on lecithinase and lipase production by Clostridium spp1
  • Ready-to-use – Convenience of prepared media
Clostridium spp. produce the enzymes lecithinase and lipase. Egg yolk suspension (50%) present in the medium serves as the substrate to detect lecithinase and lipase activity3. Lecithinase degrades lecithin and produces an insoluble, opaque precipitate in the medium surrounding growth4. Lipase breaks down free fats in the egg yolk forming in an iridescent sheen on the colony surface.
This medium contains gelatin peptone that supplies amino acids and other nitrogenous compounds necessary for the growth of anaerobic bacteria. Dextrose serves as an energy source. Egg yolk serves as a substrate for detection of lecithinase and lipase activity3 and also reduces the toxic effect of organic peroxides which may accumulate in the medium. Neomycin acts as the selective agent which inhibits many gram-positive and gram-negative organisms. Agar is the solidifying agent.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description McClung-Toabe Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.