Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mannitol Egg Yolk Polymyxin Agar

Catalog No. R01584 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R01584 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R01584 Supplier Thermo Scientific™ Supplier No. R01584
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate Bacillus spp. from foods using Mannitol Egg Yolk Polymyxin Agar.

Cultivate and enumerate Bacillus cereus in foods using Thermo Scientific™ Remel™ Mannitol Egg Yolk Polymyxin Agar. It relies on the selective inhibitory component polymyxin and two indicator systems, mannitol and phenol red and egg yolk. Thus, the growth of many unwanted organisms is suppressed. It is recommended by the Association of Official Analytical Chemists (AOAC) and the American Public Health Association (APHA).1-3

Food Poisoning by Bacillus cereus may occur when foods have been prepared and held without adequate refrigeration before being served. B. cereus is an adequate spore former which is commonly found in vegetables, many raw and processed foods, meat, cereals, pasteurized fresh milk and powdered milk. This organism causes gastrointestinal illness4.

Use Mannitol Egg Yolk Polymyxin Agar for cultivation and enumeration of Bacillus cereus in foods.

  • Easy-to-differentiate: Acid produced in the fermentation of mannitol by Bacillus spp other that B. cereus causes the phenol red indicator to change the colony color from pink to yellow.
  • Selective: Polymyxin B is a selective agent which inhibits many gram-negative bacilli and gram-positive cocci.
  • Recommended by AOAC and APHA: Recommended by the Association of Official Analytical Chemists (AOAC) and the American Public Health Association (APHA).1-3
  • Ready-to-use: Convenience of prepared media.

This medium contains beef extract and peptones that supply nitrogen, amino acids, and peptides necessary for bacterial growth. Mannitol is the carbon source. Acid produced in the fermentation of mannitol by Bacillus spp other that B. cereus causes the phenol red indicator to change the colony color from pink to yellow. B. cereus does not ferment mannitol and produces pink colonies on MEP Agar. Egg yolk supplies lecithin  and serves to reduce the toxic effect of organic peroxides which may accumulate in the agar. Lecithinase, an enzyme produced by B. cereus, metabolizes lecithin and produces an insoluble, opaque precipitation in the medium surrounding growth. Polymyxin B is a selective agent which inhibits many gram-negative bacilli and gram-positive cocci.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM.
Description MEP Agar (Mannitol, Egg Yolk, Polymyxin Agar)
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.