Learn More
Thermo Scientific™ Methanol
Catalog No. r40121
Click to view available options
Quantity:
Each
This item is not returnable. View return policy
This item is not returnable. View return policy
Absolute methanol is used with the modified acid-fast stain procedure to detect Cryptosporidium in clinical specimens.
Thermo Scientific™ Remel™ Absolute Methanol is used with the modified acid-fast stain procedure for detection of Cryptosporidium in clinical specimens. Ma and Soave recommended a modified acid-fast staining procedure because of its simplicity and specificity for detecting Cryptosporidium1. Garcia recommended a more recent modification of the Kinyoun carbolfuchsin stain that is used to detect Cryptosporidium oocysts in clinical specimens 2,3. Absolute Methanol is a fixative used to preserve the morphology of the cells and aids in detection of Cryptosporidium oocysts.
Cryptosporidium is an intestinal protozoan parasite well known for causing diarrhoea in animals and has more recently been recognized for causing human disease. Oocysts are spherical 4-6μm in diameter, and stain pink to red to purple with Kinyoun carbolfuchsin stain. Most will contain several dark staining granules.
  • Ready-to-use – Prepared reagents
  • Easy-to-store – Store in its original container at room temperature (20-25°C) until further use
  • Easy to interpret – Oocysts are spherical and stain pink to red to purple with Kinyoun carbolfuchsin stain whereas yeast cells which are morphologically like Cryptosporidium, stain blue or green depending on the counterstain used.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Type Methanol Fixative
Quantity Each
Volume (Metric) 250 mL/Bottle
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.