Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MIL Medium (Motility Indole Lysine Medium)

Catalog No. r061350
Click to view available options
Quantity:
20 x 5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Differentiate enteric gram-negative bacilli using MIL Medium.

Differentiate enteric gram-negative bacilli using Thermo Scientific™ Remel™ MIL Medium based on motility, production of indole, lysine deamination and lysine decarboxylation. MIL Medium was described by Reller and Mirrett in 1975 to facilitate early recognition and presumptive identification of members of the family Enterobacteriaceae1.

Differentiate enteric gram-negative bacilli based on motility, production of indole, lysine deamination and lysine decarboxylation using MIL Medium.

  • Easy-to-differentiate: Brom cresol purple is a pH indicator. Lysine decarboxylase activity shows enteric bacteria producing purple color in the butt of the tube. Lysine deaminase activity shows a deep red band on the rim of the medium and yellow color in the butt of tube. Indole activity shows development of pink to red color in the upper layer of the medium after addition of Kovacs reagent.
  • Ready-to-use: Convenience of prepared media.

This medium contains peptones and tryptone that supply nitrogenous compounds and amino acids necessary for enteric gram-negative bacilli. Yeast extract is a source of B-complex vitamins and dextrose is an energy source. Agar is added to enable detection of motility. Brom cresol purple is a pH indicator that is purple at the initial pH of the medium. Enteric gram-negative bacilli ferment dextrose, producing acid which changes the pH indicator color to yellow and stimulates enzyme activity. Lysine is decarboxylated by some enteric bacteria into cadaverine. This produces a purple butt in the tube because the pH indicator reverts to purple. Lysine deamination produces α-ketoglutaric acid that results in a deep red rim at the surface of the medium and a yellow butt in the tube. Indole is produced by organisms that possess the enzyme tryptophanase which degrades tryptophan present in the medium. This reacts with Kovas reagent and forms a red complex.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description MIL Medium (Motility Indole Lysine Medium)
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.