Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Mono-Lex System
Click to view available options
Quantity:
100 Tests
1000 Tests
Description
Thermo Scientific™ Remel Mono-Lex System is a rapid latex agglutination test used for detection of heterophile (Paul-Bunnell) antibodies associated with infectious mononucleosis in human serum or plasma.

Specifications
Specifications
Content And Storage | 2°C to 8°C |
Description | Mono-Lex System |
Detectable Analytes | Mononucleosis Antibodies |
Sample Type | Plasma, Serum |
Type | Reagent |
Quantity | 1000 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction