Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Motility Test Medium w/TTC

Catalog No. p-4524202
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Perform motility testing in microorganisms using Motility Test Medium w/TTC Indicator.

Demonstrate motility in microorganisms with Thermo Scientific™ Remel™ Motility Test Medium w/TTC Indicator.

Motility Test Medium w/TTC Indicator is a modification of the formulation of Tittsler and Sandholzer for detection of motility in enteric gram-negative bacilli1. When results of motility determinations in hanging-drop preparations and semi-solid agar were compared, they observed perfect correlation2. Kelly and Fulton recommended the addition of a tetrazolium salt to enhance the macroscopic appearance of motile organisms in motility media3.

  • Tetrazolium dye added to aid in detection of motility
TRUSTED_SUSTAINABILITY

Specifications

Description Motility Test Medium w/TTC
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.