Learn More
Thermo Scientific™ Motility Test Medium
Description
Perform motility testing of microorganisms using Motility Test Medium.
Demonstrate motility in microorganisms with Thermo Scientific™ Remel™ Motility Test Medium.
Motility Test Medium is a modification of the formulation of Tittsler and Sandholzer for detection of motility in enteric gram-negative bacilli1. When results of motility determinations in hanging-drop preparations and semi-solid agar were compared, they observed perfect correlation2.
Motility Test Medium is a semisolid media that is used to demonstrate motility in microorganisms. Agar provides a semisolid medium which allows macroscopic motility interpretations.
- Ready-to-use: Convenience of prepared media.
- Easy to interpret: Based on motility.
Motility Test Medium contains gelatin peptone and beef extract that supply vitamins and nitrogenous compounds essential for bacterial growth. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AOAC, COMPF. |
| Description | Motility Test Medium |
| Format | 15mm x 103mm Tube |
| Product Type | Microbiology Media |
| Quantity | 100 x 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.