Learn More
Thermo Scientific™ Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide
Catalog No. p-4523324
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
1 oz. Bottle
20mm x 113mm Tube
Monoplate Deep Pour
This item is not returnable. View return policy
This item is not returnable. View return policy
Isolate and cultivate pathogenic fungi from clinical specimens which may be contaminated with bacteria and fungi with this selective medium.
Isolate and cultivate pathogenic fungi, especially from clinical specimens which may be contaminated with bacteria and saprophytic fungi with Thermo Scientific™ Remel™ Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide. Leach, Ford and Whiffen described the use of cycloheximide as a selective agent for the inhibition of saprophytic fungi1. Researchers later found that the addition of both cycloheximide and chloramphenicol achieved more complete selectivity against growth of saprophytic fungi and bacteria2.
The Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide is recommended for recovery of dimorphic fungi and dermatophytes potentially contaminated with bacteria and saprophytic fungi3,4.
  • Selective: chloramphenicol and cycloheximide in the medium act as selective agents. Cycloheximide inhibits rapidly growing saprophytic fungi that may overgrow slower growing pathogens. Chloramphenicaol is a broad-spectrum antibiotic which inhibits Gram-positive and Gram-negative bacteria as well as Nacardia.
  • Ready prepared: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
.
TRUSTED_SUSTAINABILITY
Specifications
Description Mycobiotic Agar (Mycocel Agar) w/chloramphenicol, cycloheximide
Format Monoplate Deep Pour
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.