Learn More
Description
Use NET Test for rapid presumptive identification of Neisseria gonorrhoeae, Neisseria meningitidis, Neisseria lactamica, and Moraxella catarrhalis.
Use Thermo Scientific™ Remel™ NET Test for the rapid, presumptive identification of Neisseria gonorrhoeae, Neisseria meningitidis, Neisseria lactamica and Moraxella catarrhalis. In 1978, D'Amato et al. described a method to identify N. gonorrhoeae and N. meningitidis using chromogenic substrates to determine enzyme profiles1. Hoke et al. used modification of his technique to characterize atypical strains of N. gonorrhoeae and N. meningitidis2. In 1982, Yajko et al. described a method to identify Neisseria spp. using mixed chromogenic substrates3.
NET employs three synthetic chromogenic substrates to detect preformed enzymes. The enzyme β-D-galactopyranoside forms a blue colored complex in the presence of oxygen and γ-glutamyl peptidase hydrolyzes γ-glutamyl-ρ-nitroanilide to form a yellow color. The third enzyme, prolylaminopeptidase, hydrolyzes L-proline β-naphthylamide which is detected by a color change to pink-red after adding ρ-dimethyl-aminocinnamaldehyde.
- Ready to use – Prepared reagents.
- Easy to store – Store in its original container at room temperature (2-8°C) until further use.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | NET |
| Includes | Lyophilized reagent in tubes; phosphate buffered saline; and PRO reagent |
| Quantity | 25 Tests/Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
