Learn More
Description
Determine the ability of an organism to reduce nitrate to nitrite or free nitrogen gas using Thermo Scientific™ Remel™ Nitrate Reagent B. This ability is characteristic of the Enterobacteriaceae1 family. Nonfermenters and other miscellaneous gram-negative bacilli vary in their ability to reduce nitrates, with some being capable of further reducing nitrite to nitrogen gas2. In 1962, Daubner reported that all members of the Enterobacteriaceae tested, except for Erwinia species, reducing nitrate in eight hours3.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Nitrate Reagent B |
| Detectable Analytes | Nitrite, Free Nitrogen Gas |
| Format | Bottle |
| Quantity | 25 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
