Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ OF Medium w/1% Dextrose

Catalog No. r061916
Click to view available options
Quantity:
20 x 4 mL
100 x 4 mL
This item is not returnable. View return policy

Catalog No. R061916

Supplier: Thermo Scientific™ R061916

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use OF Medium w/1% Dextrose (Hugh-Liefson) to determine oxidative and fermentative metabolism of carbohydrates by gram-negative bacilli.

Determine oxidative and fermentative metabolism of glucose by gram-negative bacilli using Thermo Scientific™ Remel™ OF Medium w/1% Dextrose (Hugh-Liefson). Hugh and Leifson first reported that microorganisms which utilize carbohydrates oxidatively produce only a minimal amount of acid1. Oxidative-Fermentative (OF) Medium was developed with a ratio of peptone to carbohydrate that minimized the formation of alkaline by-products and enhanced acid production by oxidative organisms. OF Medium is especially suitable for determining the carbohydrate metabolic pathway of gram-negative bacilli2.

Typical fermentation media contain approximately 2% peptone and 1% carbohydrate. In media containing high concentrations of peptones, oxidative microorganisms produce alkaline by-products in quantities that neutralize the acid produced from carbohydrate metabolism.

OF Medium is a semisolid medium with a low peptone to carbohydrate ratio that contains the pH indicator bromthymol blue. Organisms which metabolize carbohydrate in OF medium produce acid in quantities sufficient to prevent its neutralization by alkaline peptone by-products.

  • Ready to use – Convenience of prepared media
  • Uses 4 mL of 1% Dextrose as carbohydrate source
  • Easy to interpret – Color change from green to yellow indicates positive test for acid production.
    When testing for motility, growth diffused away from stab line of inoculation indicates a positive test whereas growth confined to stab line indicates negative test.
    Acid production in the open tube (without mineral oil overlay) indicates oxidative metabolism whereas acid production in both (with and without mineral oil overlay) tubes indicates fermentative metabolism

OF Medium with Dextrose uses 4 mL of 1% dextrose as a fermentable carbohydrate. The medium contains casein peptone as the substrate and bromthymol blue as the pH indicator. Dipotassium phosphate promotes fermentation and also serves as a buffer. Agar allows for detection of motility and aids in even distribution of acid produced at the surface of the medium.

Dextrose is routinely used in OF Medium; however, some organisms may be unable to metabolize dextrose but can utilize other carbohydrates. Organisms that only oxidise glucose will not ferment any other carbohydrate1. Therefore, when testing with other carbohydrates, the mineral oil overlay tube may be omitted.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM.
Description OF Medium w/1% Dextrose (Hugh-Liefson)
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 100 x 4 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.