Learn More
Thermo Scientific™ Phenylethyl Alcohol Agar with 5% Sheep Blood
Description
Isolate and cultivate Gram-positive cocci, such as staphylococci and streptococci, from specimens containing mixed flora1 with Thermo Scientific™ Remel™ Phenylethyl Alcohol Agar with 5% Sheep Blood. This medium permits the growth of Gram-positive organisms while inhibiting most Gram-negative organisms, especially the swarming of Proteus species.
Phenylethyl Alcohol Agar with 5% Sheep Blood is formulated as per Lilley and Brewer1 for selective isolation of Gram-positive bacteria. This medium is especially useful when specimens are contaminated with swarming Proteus species.
- Selective: phenylethyl alcohol in the medium inhibits Gram-negative organisms by inhibiting their DNA synthesis.
- Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Phenylethyl Alcohol Agar w/5% Sheep Blood |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar |
| Quantity | 10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.