Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Phosphate Buffer

Catalog No. p-4528592
Click to view available options
Quantity:
50 mL
250 mL
500 mL
This item is not returnable. View return policy

Catalog No. R21256

Supplier: Thermo Scientific™ R21256

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel Phosphate Buffer is for neutralization of clinical specimens in digestion and decontamination procedures for AFB and fungal cultures.

Thermo Scientific™ Remel Phosphate Buffer is for neutralization of clinical specimens in digestion and decontamination procedures for AFB and fungal cultures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Phosphate Buffer
Format Bottle
Quantity 50 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.