Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phosphate Buffered Saline (Cold Enrichment Broth) pH 7.6

Catalog No. R062580
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Selectively isolate Yersinia enterocolitica by cold enrichment method using Phosphate Buffered Saline (Cold Enrichment Broth) pH 7.6.

Selectively isolate Yersinia enterocolitica using Thermo Scientific™ Remel™ Phosphate Buffered Saline (Cold Enrichment Broth) pH 7.6. Cold temperature enrichment of clinical specimens prior to inoculation of media has been reported to improve recovery of Y. enterocolitica from fecal specimens1-3. Weissfeld and Sonnenwirth demonstrated cold enrichment of fecal specimens resulted in an incidence rate of yersiniosis equal to that of salmonellosis4.

Yersinia enterocolitica is challenging to isolate and identify in the clinical laboratory because it grows slowly at 37°C and produces biochemical reactions that are very similar to other enteric gram-negative bacilli5. Y. enterocolitica can survive and grow at the low temperature, while most other microorganisms cannot. The ability of Y. enterocolitica to grow at cold enrichment temperatures to improve the recovery of certain bacteria from foods, environmental samples and clinical specimens from asymptomatic carriers and patients who are beyond the acute (diarrheal) stage of illness2,3,6-10.

  • Ready to use: Convenience of a prepared media
  • Selective: Phosphate Buffered Saline pH 7.6, when used at refrigeration temperatures, acts as a selective medium for Yersinia in food and clinical specimens
  • Useful for epidemiological studies when specimens are contaminated with commensal microbial flora

The medium contains sodium chloride which maintains osmotic equilibrium. Monosodium and disodium phosphate salts act as buffers to maintain the pH at 7.6 ± 0.2 @25° C.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Phosphate Buffered Saline
Format 15mm x 103mm Tube
Product Type Buffer Solution
Quantity 100 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.