Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Potassium Chloride Solution

Catalog No. p-4523986
Click to view available options
Quantity:
20 x 4.5 mL
100 x 4.5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium Chloride Solution (pH 2.2, 0.2M) is used as a pretreatment to increase yield of Legionella from clinical specimens.

Pretreat the samples to increase yield of Legionella from clinical specimens using Thermo Scientific™ Remel™ Potassium Chloride Solution (pH 2.2, 0.2M). In 1981, Bopp et al. demonstrated the recovery rate of Legionella from environmental water samples was improved by treating samples with acid prior to plating onto selective media1. Buesching et al. demonstrated the recovery of Legionella organisms from clinical specimens was improved by pre-treatment with potassium chloride-hydrochloric acid solution (pH 2.2)2.

Potassium Chloride Solution (pH 2.2, 0.2M) is a liquid medium that is used as a pretreatment to increase yield of Legionella from clinical specimens. Potassium Chloride Solution (pH 2.2, 0.2M) is an acid wash treatment which serves to decontam

TRUSTED_SUSTAINABILITY

Specifications

Description Potassium Chloride Solution
Format 15mm x 103mm Tube
Product Type Buffer Solution
Quantity 100 x 4.5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.