Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Potato Dextrose Agar, pH 5.6

Catalog No. r112535
Click to view available options
Quantity:
10 x 500 mL
10/Pk.
100/Pk.
20/Pk.
Format:
BB - 500mL
F - slant
G - PT
Monoplate Deep Pour
This item is not returnable. View return policy
This item is not returnable. View return policy

Identify, cultivate and enumerate yeast and molds with this medium.

Identify, cultivate and enumerate yeast and molds with Thermo Scientific™ Remel™ Potato Dextrose Agar. Recommended by American Public Health Association (APHA) and Association of Official Analytical Chemists (AOAC), for plate count of yeasts and molds in the inspection of dairy products and foods1,2,3. This medium is used to prepare slide culture of fungi to stimulate the spores4,5. It is used to maintain stock cultures of dermatophytes. Rebel and Taplin used this medium to differentiate atypical dermatophytes based on the pigment produced6.

Identify, cultivate and enumerate yeast and molds using the solid medium Potato Dextrose Agar with pH 5.6.

  • Recommended by APHA and AOAC, for plate count of yeasts and molds in the inspection of dairy products and foods1,2,3.
  • <
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF, USP.
Description Potato Dextrose Agar pH 5.6
Format BB - 500mL
Product Type Agar
Quantity 10 x 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.