Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pseudomonas F Agar

Catalog No. p-4523300
Click to view available options
Quantity:
10/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Detect and differentiate Pseudomonas aeruginosa from other Pseudomonas spp. based on fluorescein production using Pseudomonas F Agar.

Detect and differentiate Pseudomonas aeruginosa from other Pseudomonas spp. based on fluorescein production using Thermo Scientific™ Remel™ Pseudomonas F Agar. In 1954, King et al. developed two media formulations (Medium A and Medium B) to enhance pigment production in pseudomonads1. Earlier work had demonstrated that the production of pyocyanin and fluorescein is influenced by amino acids, inorganic ions, minerals, and peptones. King determined that the composition of the peptone in the medium affected the production of these pigments.

Pseudomonas F Agar is used for the differentiation of Pseudomonas aeruginosa from other pseudomonads based on fluorescein production. Pseudomonas F Agar was developed by King, Ward, and Raney to give improved fluorescein production. The absenc

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, USP.
Description Pseudomonas F Agar (FLO Agar)
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.