Learn More
Thermo Scientific™ R2A Agar
Catalog No. r110722
Click to view available options
Quantity:
10 x 200 mL
10 x 500 mL
10/Pk.
100/Pk.
Format:
15mm x 60mm Plate
BB - 200mL
BB - 500mL
Monoplate
This item is not returnable. View return policy
This item is not returnable. View return policy
Enumerate and cultivate heterotrophic bacteria from potable water using R2A Agar.
Enumerate and cultivate heterotrophic bacteria from potable water using Thermo Scientific™ Remel™ R2A Agar. The growth of stressed and chlorine-tolerant bacteria recovered from treated waters is stimulated using a less nutritious medium, lower incubation temperatures, and longer incubation times. R2A Agar is recommended by the American Public Health Association (APHA) in Standard Methods for the Examination of Water and Wasterwater1. Information on water quality, such as heterotrophic plate counts, may be used to evaluate the efficiency of treatment processes.
Use R2A Agar for enumeration and cultivation of heterotrophic bacteria from potable water.
  • Recommended by the APHA in Standard Methods for the Examination of Water and Wasterwater1
  • Superior to plate count agar whe
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: SMWW.
Description R2A Agar
Format Monoplate
Product Type Agar
Quantity 100/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.