Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ R2A Agar

Catalog No. r112542
Click to view available options
Quantity:
10 x 200 mL
10 x 500 mL
10/Pk.
100/Pk.
Format:
15mm x 60mm Plate
BB - 200mL
BB - 500mL
Monoplate
This item is not returnable. View return policy
This item is not returnable. View return policy

Enumerate and cultivate heterotrophic bacteria from potable water using R2A Agar.

Enumerate and cultivate heterotrophic bacteria from potable water using Thermo Scientific™ Remel™ R2A Agar. The growth of stressed and chlorine-tolerant bacteria recovered from treated waters is stimulated using a less nutritious medium, lower incubation temperatures, and longer incubation times. R2A Agar is recommended by the American Public Health Association (APHA) in Standard Methods for the Examination of Water and Wasterwater1. Information on water quality, such as heterotrophic plate counts, may be used to evaluate the efficiency of treatment processes.

Use R2A Agar for enumeration and cultivation of heterotrophic bacteria from potable water.

  • Recommended by the APHA in Standard Methods for the Examination of Water and Wasterwater1
  • Superior to plate count agar whe
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: SMWW.
Description R2A Agar
Format BB - 200mL
Product Type Agar
Quantity 10 x 200 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.