Learn More
Thermo Scientific™ Remel™ Biplates: Anaerobic Reducible through Levine Agars
Click to view available options
Quantity:
10/Pk.
100/Pk.
Media Type:
Chocolate /Thayer Martin Agar
Columbia CNA/MacConkey Agar
Description
Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel™ Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.
Specifications
Specifications
| Format | Biplate |
| Media Type | Columbia CNA/MacConkey Agar |
| Product Type | Agar |
| Quantity | 100/Pk. |