Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SABHI™ Agar (Sabouraud Dextrose with Brain Heart Infusion Agar)

Catalog No. r08692
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
20mm x 113mm Tube
Monoplate Deep Pour
This item is not returnable. View return policy
This item is not returnable. View return policy

Cultivate and isolate pathogenic fungi with this selective, ready-to-use medium.

Cultivate and isolate pathogenic fungi with Thermo Scientific™ Remel™ SABHI™ Agar. Sabouraud Dextrose Agar was developed by Sabouraud for cultivation of dermatophytes1. Brain Heart Infusion Agar is useful for primary isolation of fungi from clinical specimens2. In 1967, Gorman developed SABHI Agar combining Sabouraud Dextrose Agar and Brain Heart Infusion Agar. This combined formulation was found to increase the recovery of pathogenic fungi.

Pathogenic fungi have an enormous impact on human health. Most people are aware of some of the superficial infections caused by fungi. These include skin and nail infections such as athlete's foot and ringworm, predominantly caused by a dermatophyte. More seriously, certain fungal species can cause invasive diseases in humans.

Use SABHI™ Agar for the cultivation and isolation of pathogenic fungi.

  • The combined formulation of Brain Heart Infusion Agar and Sabouraud Dextrose Agar gives improved recovery of pathogenic fungi.
  • Ready-to-use convenience of prepared media.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description SABHI Agar
Format 20mm x 113mm Tube
Product Type Agar
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.