Learn More
Thermo Scientific™ Remel™ Sabouraud Dextrose Agar Bactiflask™
Click to view available options
Quantity:
20/Pk.
Description
Thermo Scientific™ Remel Sabouraud Dextrose Agar Bactiflask (pH 5.6) is used for cultivation and isolation of yeast and fungi.
Lower pH enhances growth of fungi, especially dermatophytes and is slightly inhibitory to bacteria.
Specifications
Specifications
Description | Sabouraud Dextrose Agar |
Format | Bactiflask - 1 oz. |
Product Type | Agar |
Quantity | 20/Pk. |