Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sabouraud Dextrose Agar (Emmons)

Catalog No. r064420
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube
15mm x 150mm Plate
20mm x 113mm Tube
Monoplate Deep Pour
This item is not returnable. View return policy
This item is not returnable. View return policy

Isolate and cultivate pathogenic yeast and molds from clinical specimens with this selective medium.

Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar (Emmons) prepared media plates. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concertation of dextrose from 40 to 20 g/L2.

Use Sabouraud Dextrose Agar (Emmons) for primary isolation and cultivation of pathogenic yeasts and molds from clinical specimens.

  • Suitable for isolation of pathogenic fungi from a material containing a large amount of other fungi or bacteria.
  • Ready prepared convenience of prepared media.

Casein and meat peptones supply nitrogenous compounds and amino acids necessary for the growth of fungi. Dextrose is an energy source which serves to support the growth of fungi. Chloramphenicol and gentamicin are broad-spectrum antibiotics which inhibit a range of Gram-positive and Gram-negative bacteria. Cycloheximide is inhibitory to rapidly growing saprophytic fungi that could overgrow slow-growing primary pathogens. Penicillin is active against Gram-positive bacteria. Streptomycin inhibits Gram-positive and Gram-negative bacteria.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Sabouraud Dextrose Agar, Emmons
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 100/Pk.

REF R112555 not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.