Learn More
Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, Emmons w/Chloramphenicol Bactiflask™
Click to view available options
Quantity:
20/Pk.
Format:
1 oz. TCF
Description
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons w/Chloramphenicol Bactiflask is used for selective isolation of yeast and fungi.
Chloramphenicol inhibits some gram-positive and gram-negative bacteria.
Specifications
Specifications
Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol Bactiflask™ |
Format | 1 oz. TCF |
Product Type | Agar |
Quantity | 20/Pk. |