Learn More
Thermo Scientific™ Sabouraud Dextrose Agar, Emmons w/Chloramphenicol
Catalog No. p-4522759
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
F - slant
Monoplate Deep Pour
This item is not returnable. View return policy
This item is not returnable. View return policy
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20 g/l2. Chloramphenicol is added to the medium to make it inhibitory to bacterial overgrowth while allowing isolation of pathogenic fungi3.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol.
  • Ready-to-use – Convenience of a prepared media.
  • Suitable for isolation of pathogenic fungi from a material containing a large number of other fungi or bacteria.
  • Selective – Chloramphenicol, which is a broad-spectrum antibiotic, inhibits the growth of a range of gram-positive and gram-negative bacteria.
This medium contains casein, meat peptones, nitrogenous compounds and amino acids necessary for the growth of fungi. Dextrose is an energy source which serves to support the growth of fungi. Chloramphenicol is a broad-spectrum antibiotic which inhibit a range of Gram-positive and Gram-negative bacteria. Yeast extract may be added to the medium as a growth supplement to ensure the growth of fastidious fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Sabouraud Dextrose Agar, Emmons w/Chloramphenicol
Format Monoplate Deep Pour
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.