Learn More
Thermo Scientific™ Sabouraud Dextrose Agar, Emmons w/Yeast Extract
Catalog No. r01780
This item is not returnable. View return policy
Catalog No. R01780
Supplier: Thermo Scientific™ R01780
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Yeast Extract.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, Emmons, with Yeast Extract. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20g/L2. Yeast extract is added to the medium as a growth supplement.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Yeast Extract.
  • Ready-to-use – Convenience of a prepared media.
  • Suitable for isolation of pathogenic fungi from a material containing a large number of other fungi or bacteria.
  • Yeast extract acts as a growth supplement for fastidious fungi.
This medium also contains casein and meat peptones which supply nitrogenous compounds and amino acids necessary for the growth of fungi. Dextrose is an energy source which serves to support the growth of fungi. Yeast extract is added to the medium as a growth supplement to ensure the growth of fastidious fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Sabouraud Dextrose Agar, Emmons w/Yeast Extract
Format Monoplate Deep Pour
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.