Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Salmonella Shigella Agar (SS Agar)

Catalog No. r01842
Click to view available options
Quantity:
10/Pk.
100/Pk.
Format:
12mm x 85mm Monoplate
This item is not returnable. View return policy
This item is not returnable. View return policy

Use for the selective and differential isolation of Salmonella and to a lesser degree, Shigella spp.

Thermo Scientific™ Remel Salmonella Shigella Agar (SS Agar) is used for the selective and differential isolation of Salmonella and to a lesser degree, Shigella spp.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: COMPF, USDA.
Description Salmonella Shigella (SS) Agar
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 100/Pk.

Part No. R119756 not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.