Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ SP4 Glucose Broth

Catalog No. r20376
Click to view available options
Format:
CSB - 100mL
CSB - 90mL
D - 5mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo Scientific™ Remel SP4 Glucose Broth (w/thallium acetate, penicillin) is used for isolation of Mycoplasma species.

Thermo Scientific™ Remel SP4 Glucose Broth (w/thallium acetate, penicillin) is used for isolation of Mycoplasma species.

  • Penicillin inhibits gram-positive bacteria
TRUSTED_SUSTAINABILITY

Specifications

Description SP4 Glucose Broth w/thallium acetate, penicillin
Format D - 5mL
Product Type Microbiology Media
Quantity 20 x 5 mL

REF R112585 not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.