Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Starch Hydrolysis Agar

Catalog No. R01854
Click to view available options
Quantity:
10/Pk.
Format:
12mm x 85mm Monoplate
This item is not returnable. View return policy

Catalog No. R01854

Supplier: Thermo Scientific™ R01854

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Detect amylolytic activity or hydrolysis of starch by a wide variety of organisms using Starch Hydrolysis Agar.

Detect amylolytic activity or hydrolysis of starch by organisms with Thermo Scientific™ Remel™ Starch Hydrolysis Agar. Starch Agar was originally formulated by Vedder in the year 1915 for the cultivation of Neisseria1. Heart Infusion Agar is used as the base for Starch Hydrolysis Agar with the addition of soluble starch2. This medium is used for differentiation of Clostridium perfringens (positive) from other Clostridium spp.3,4

Starch Hydrolysis Agar is a solid medium recommended for use in qualitative procedures to detect amylolytic activity or hydrolysis of starch by a wide variety of organisms.

Starch acts as the substrate which the organism may hydrolyze by producing amylase.

  • Ready-to-use: Convenience of prepared media.
  • Easy to interpret: A colorless zone around the colonies indicates a positive test. Blue color around the colonies indicates a negative result.

Starch Hydrolysis Agar contains beef heart infusion and casein peptone that supply nutrients necessary for growth of all except the very fastidious bacteria. Yeast extract acts as a source of energy and procides additional nutritional requirements. Sodium chloride supplies essential electrolytes and maintains osmotic equilibrium. Starch is the fermentable substrate.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Starch Hydrolysis Agar
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.